Assay Details
Target Gene Details
Entrez Gene ID: | 7412 |
Gene Name: | vascular cell adhesion molecule 1 |
Gene Aliases: |
CD106, INCAM-100 |
Location: |
Chr.1:100719640-100739045 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCAM1 | NM_001078.3 | 2 | 340 | NP_001069.1 |
| NM_001199834.1 | 2 | 340 | NP_001186763.1 | |
| NM_080682.2 | 2 | 340 | NP_542413.1 | |
| AK223266.1 | 2 | 239 | BAD96986.1 | |
| AK291732.1 | 2 | 238 | ||
| AK296692.1 | 2 | 238 | ||
| AU121762.1 | 2 | 240 | ||
| BC017276.2 | 2 | 220 | AAH17276.3 | |
| BC068490.1 | 2 | 238 | AAH68490.2 | |
| BC085003.1 | 2 | 220 | AAH85003.1 | |
| DB096825.1 | 2 | 340 | ||
| M30257.1 | 2 | 225 | AAA51917.1 | |
| M60335.1 | 2 | 218 | AAA61269.1 | |
| X53051.1 | 2 | 119 | CAA37218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
| nsv546889 | Chr.1:100504790 - 100725310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map