Assay Details
Target Gene Details
Entrez Gene ID: | 148523 |
Gene Name: | circadian associated repressor of transcription |
Gene Aliases: |
C1orf51, CHRONO, GM129 |
Location: |
Chr.1:150282533-150287093 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIART | NM_001300838.1 | NP_001287767.1 | ||
| NM_001300839.1 | NP_001287768.1 | |||
| NM_001300840.1 | NP_001287769.1 | |||
| NM_001300841.1 | NP_001287770.1 | |||
| NM_144697.3 | 1 | 381 | NP_653298.1 | |
| XM_017000378.1 | XP_016855867.1 | |||
| AK098755.1 | 1 | 65 | BAC05404.1 | |
| AK315397.1 | 1 | 65 | ||
| BC017397.1 | AAH17397.1 | |||
| BC027999.1 | AAH27999.1 | |||
| BC047238.1 | 1 | 95 | AAH47238.1 | |
| BE784098.1 | ||||
| CN359562.1 | ||||
| EB388133.1 | 1 | 381 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009508 | Chr.1:150238763 - 150298405 on Build GRCh38 | Loss |
|
| nsv1011223 | Chr.1:150256810 - 150306949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map