Assay Details
Target Gene Details
Entrez Gene ID: | 2059 |
Gene Name: | epidermal growth factor receptor pathway substrate 8 |
Gene Aliases: |
DFNB102 |
Location: |
Chr.12:15620141-15789576 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS8 | NM_004447.5 | 14 | 1783 | NP_004438.3 |
| XM_011520605.2 | 15 | 1843 | XP_011518907.1 | |
| AK291777.1 | 14 | 1588 | ||
| AK292931.1 | 14 | 1440 | ||
| AK301834.1 | 7 | 773 | ||
| AK304860.1 | 14 | 1783 | ||
| AK316134.1 | 7 | 692 | ||
| AK316239.1 | 7 | 965 | ||
| BC030010.2 | 14 | 1595 | AAH30010.1 | |
| U12535.1 | 14 | 1555 | AAA62280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760899 | Chr.12:15643377 - 15943563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map