Assay Details
Target Gene Details
Entrez Gene ID: | 9783 |
Gene Name: | regulating synaptic membrane exocytosis 3 |
Gene Aliases: |
NIM3, RIM3 |
Location: |
Chr.1:40620680-40692104 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMS3 | NM_014747.2 | 8 | 2142 | NP_055562.2 |
| XM_011542479.2 | 7 | 1780 | XP_011540781.1 | |
| D87074.1 | 8 | 2148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546064 | Chr.1:40431820 - 40673669 on Build GRCh38 | Gain |
|
| nsv599 | Chr.1:40614896 - 40648261 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map