Assay Details
Target Gene Details
Entrez Gene ID: | 1806 |
Gene Name: | dihydropyrimidine dehydrogenase |
Gene Aliases: |
DHP, DHPDHASE, DPD |
Location: |
Chr.1:97077743-97921059 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD | NM_000110.3 | 23 | 3805 | NP_000101.2 |
| XM_005270562.3 | 22 | 3589 | XP_005270619.2 | |
| XM_017000507.1 | 22 | 3688 | XP_016855996.1 | |
| XM_017000508.1 | 25 | 4015 | XP_016855997.1 | |
| XM_017000509.1 | 24 | 3913 | XP_016855998.1 | |
| XM_017000510.1 | 23 | 3802 | XP_016855999.1 | |
| U09178.1 | 23 | 3749 | AAA57474.1 | |
| U20938.1 | 23 | 3769 | AAB51366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830692 | Chr.1:97035936 - 97159597 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map