Assay Details
Target Gene Details
Entrez Gene ID: | 7292 |
Gene Name: | tumor necrosis factor superfamily member 4 |
Gene Aliases: |
CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1 |
Location: |
Chr.1:173183729-173462208 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF4 | NM_001297562.1 | 3 | 515 | NP_001284491.1 |
| NM_003326.4 | 3 | 557 | NP_003317.1 | |
| XM_011509964.2 | 4 | 1360 | XP_011508266.2 | |
| XM_017002228.1 | 3 | 1789 | XP_016857717.1 | |
| XM_017002229.1 | 5 | 6680 | XP_016857718.1 | |
| XM_017002230.1 | 4 | 1044 | XP_016857719.1 | |
| AK297932.1 | 3 | 430 | ||
| BC041663.1 | 3 | 515 | AAH41663.1 | |
| CR542033.1 | 3 | 420 | CAG46830.1 | |
| D90224.1 | 3 | 456 | BAA14259.1 | |
| X79929.1 | 3 | 557 | CAA56284.1 |
More Information
Set Membership: |
|

Genomic Map