Assay Details
Target Gene Details
Entrez Gene ID: | 2205 |
Gene Name: | Fc fragment of IgE receptor Ia |
Gene Aliases: |
FCE1A, FcERI |
Location: |
Chr.1:159283888-159308224 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCER1A | NM_002001.3 | NP_001992.1 | ||
| BC005912.1 | AAH05912.1 | |||
| BC015195.1 | AAH15195.1 | |||
| J03605.1 | AAA36204.1 | |||
| X06948.1 | CAA30025.1 |
More Information
Set Membership: |
|

Genomic Map