Assay Details
Target Gene Details
Entrez Gene ID: | 51604 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class T |
Gene Aliases: |
CGI-06, MCAHS3, NDAP, PNH2 |
Location: |
Chr.20:45416067-45426245 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGT | NM_001184728.2 | NP_001171657.1 | ||
| NM_001184729.2 | NP_001171658.1 | |||
| NM_001184730.2 | NP_001171659.1 | |||
| NM_015937.5 | NP_057021.2 | |||
| NR_047691.1 | ||||
| NR_047692.1 | ||||
| NR_047693.1 | ||||
| NR_047694.1 | ||||
| NR_047695.1 | ||||
| XM_005260430.2 | XP_005260487.1 | |||
| XM_005260432.2 | XP_005260489.1 | |||
| AB057724.1 | BAB60854.1 | |||
| AF058295.1 | ||||
| AF132940.1 | AAD27715.1 | |||
| AK075469.1 | BAC11639.1 | |||
| AK225517.1 | ||||
| AK293294.1 | ||||
| AK293686.1 | ||||
| AK296063.1 | ||||
| AK296139.1 | ||||
| AK299013.1 | ||||
| AK301815.1 | ||||
| AK302093.1 | ||||
| AK311100.1 | ||||
| AK311416.1 | ||||
| AK311492.1 | ||||
| AK311520.1 | ||||
| AK311522.1 | ||||
| AK316358.1 | ||||
| AL121742.1 | CAB57341.1 | |||
| AY358588.1 | AAQ88951.1 | |||
| BC015022.1 | ||||
| BC110892.1 | AAI10893.1 | |||
| BC136827.1 | ||||
| BC136828.1 | ||||
| BX537612.1 | 2 | 1946 | CAD97799.1 | |
| DA037291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
| dgv4319n100 | Chr.20:45384898 - 45428463 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map