Assay Details
Target Gene Details
Entrez Gene ID: | 51768 |
Gene Name: | transmembrane 7 superfamily member 3 |
Gene Aliases: |
- |
Location: |
Chr.12:26966519-27014427 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM7SF3 | NM_016551.2 | NP_057635.1 | ||
| XM_005253391.3 | XP_005253448.1 | |||
| XM_011520690.2 | XP_011518992.1 | |||
| XM_011520691.1 | XP_011518993.1 | |||
| XM_017019463.1 | XP_016874952.1 | |||
| XM_017019464.1 | XP_016874953.1 | |||
| XM_017019465.1 | XP_016874954.1 | |||
| XM_017019466.1 | XP_016874955.1 | |||
| AB032470.1 | BAA92856.1 | |||
| AK002031.1 | BAA92046.1 | |||
| AK023085.1 | ||||
| BC005176.2 | AAH05176.1 |
More Information
Set Membership: |
|

Genomic Map