Assay Details
Target Gene Details
Entrez Gene ID: | 55245 |
Gene Name: | ubiquinol-cytochrome c reductase complex assembly factor 1 |
Gene Aliases: |
BFZB, C20orf44, CBP3, UQCC |
Location: |
Chr.20:35302566-35412142 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCC1 | NM_001184977.1 | 8 | 2204 | NP_001171906.1 |
| NM_018244.4 | 10 | 2408 | NP_060714.3 | |
| NM_199487.2 | 9 | 2330 | NP_955781.2 | |
| XM_011528877.1 | XP_011527179.1 | |||
| XM_011528878.1 | XP_011527180.1 | |||
| XM_011528879.1 | XP_011527181.1 | |||
| XM_011528880.1 | XP_011527182.1 | |||
| XM_011528881.2 | XP_011527183.1 | |||
| XM_011528882.1 | XP_011527184.1 | |||
| XM_011528883.1 | XP_011527185.1 | |||
| XM_011528884.1 | XP_011527186.1 | |||
| AF173893.1 | AAQ13640.1 | |||
| AK001712.1 | BAA91854.1 | |||
| AK022650.1 | BAB14156.1 | |||
| AK022742.1 | BAB14217.1 | |||
| AK093804.1 | ||||
| AK128252.1 | ||||
| AL122112.1 | ||||
| AY050659.1 | 10 | 2408 | AAL13118.1 | |
| BX537568.1 | CAD97785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059168 | Chr.20:35217473 - 35319358 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map