Assay Details
Target Gene Details
Entrez Gene ID: | 4504 |
Gene Name: | metallothionein 3 |
Gene Aliases: |
GIF, GIFB, GRIF, ZnMT3 |
Location: |
Chr.16:56589355-56591088 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MT3 | NM_005954.2 | 1 | 39 | NP_005945.1 |
More Information
Set Membership: |
|

Genomic Map