Assay Details
Target Gene Details
Entrez Gene ID: | 14 |
Gene Name: | angio associated migratory cell protein |
Gene Aliases: |
- |
Location: |
Chr.2:218264129-218270209 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAMP | NM_001087.4 | NP_001078.2 | ||
| NM_001302545.1 | NP_001289474.1 | |||
| AB209790.1 | BAD93027.1 | |||
| AK131047.1 | ||||
| AK225078.1 | ||||
| BC008809.2 | AAH08809.2 | |||
| BC020244.1 | AAH20244.1 | |||
| BC039866.1 | AAH39866.2 | |||
| CR456755.1 | CAG33036.1 | |||
| HY374777.1 | ||||
| M95627.1 | AAA68889.1 |
More Information
Set Membership: |
|

Genomic Map