Assay Details
Target Gene Details
Entrez Gene ID: | 8625 |
Gene Name: | regulatory factor X associated ankyrin containing protein |
Gene Aliases: |
ANKRA1, BLS, F14150_1, RFX-B |
Location: |
Chr.19:19192199-19201869 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFXANK | NM_001278727.1 | NP_001265656.1 | ||
| NM_001278728.1 | NP_001265657.1 | |||
| NM_003721.3 | NP_003712.1 | |||
| NM_134440.2 | NP_604389.1 | |||
| XM_005260134.4 | XP_005260191.1 | |||
| XM_005260135.3 | XP_005260192.1 | |||
| XM_005260136.4 | XP_005260193.1 | |||
| XM_005260137.4 | XP_005260194.1 | |||
| XM_006722930.3 | XP_006722993.1 | |||
| XM_017027415.1 | XP_016882904.1 | |||
| XM_017027416.1 | XP_016882905.1 | |||
| AF077196.1 | AAD26991.1 | |||
| AF094760.1 | AAC69883.1 | |||
| AF105427.1 | AAD17972.1 | |||
| AF105428.1 | AAD17973.1 | |||
| AW411361.1 | ||||
| BC114558.1 | AAI14559.1 | |||
| BC114563.1 | AAI14564.1 | |||
| BG740813.1 | ||||
| CR456981.1 | CAG33262.1 | |||
| CR542199.1 | CAG46996.1 | |||
| CX871218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833779 | Chr.19:19052683 - 19202282 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map