Assay Details
Target Gene Details
Entrez Gene ID: | 93627 |
Gene Name: | TBC1 domain containing kinase |
Gene Aliases: |
HSPC302, IHPRF3, TBCKL |
Location: |
Chr.4:106044317-106316704 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCK | NM_001163435.2 | 25 | 2873 | NP_001156907.1 |
| NM_001163436.2 | 25 | 2694 | NP_001156908.1 | |
| NM_001163437.2 | 25 | 2756 | NP_001156909.1 | |
| NM_001290768.1 | 26 | 2974 | NP_001277697.1 | |
| NM_033115.4 | 23 | 2684 | NP_149106.2 | |
| XM_011532417.2 | 25 | 2793 | XP_011530719.1 | |
| XM_017008846.1 | 25 | 2877 | XP_016864335.1 | |
| XM_017008847.1 | 24 | 2537 | XP_016864336.1 | |
| XM_017008848.1 | 25 | 2549 | XP_016864337.1 | |
| XM_017008849.1 | 23 | 2360 | XP_016864338.1 | |
| AB449876.1 | 24 | 2508 | ||
| AF161420.1 | 13 | 1350 | AAF28980.1 | |
| AK074305.1 | 25 | 2628 | BAB85045.1 | |
| AK098157.1 | 16 | 2161 | BAC05244.1 | |
| AK315571.1 | 25 | 2816 | ||
| BC009208.2 | 23 | 2646 | AAH09208.2 | |
| BC020853.2 | 6 | 689 | AAH20853.2 | |
| BC068496.1 | 25 | 2697 | AAH68496.1 | |
| BX647851.1 | 27 | 2918 | CAI46102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map