Assay Details
Target Gene Details
Entrez Gene ID: | 11051 |
Gene Name: | nudix hydrolase 21 |
Gene Aliases: |
CFIM25, CPSF5 |
Location: |
Chr.16:56429136-56451349 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT21 | NM_007006.2 | 2 | 423 | NP_008937.1 |
| AJ001810.1 | 2 | 276 | CAA05026.1 | |
| AK312721.1 | 2 | 426 | ||
| AL535307.3 | 2 | 423 | ||
| BC001403.2 | 2 | 384 | AAH01403.1 | |
| BX537360.1 | 2 | 427 | CAD97606.1 | |
| BX647665.1 | CAI46057.1 | |||
| CR456919.1 | 2 | 276 | CAG33200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map