Assay Details
Target Gene Details
Entrez Gene ID: | 4948 |
Gene Name: | OCA2 melanosomal transmembrane protein |
Gene Aliases: |
BEY, BEY1, BEY2, BOCA, D15S12, EYCL, EYCL2, EYCL3, HCL3, P, PED, SHEP1 |
Location: |
Chr.15:27719008-28099342 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OCA2 | NM_000275.2 | 24 | 2588 | NP_000266.2 |
| NM_001300984.1 | 23 | 2516 | NP_001287913.1 | |
| XM_011521640.2 | 25 | 2660 | XP_011519942.1 | |
| XM_017022255.1 | 25 | 2611 | XP_016877744.1 | |
| XM_017022256.1 | 24 | 3066 | XP_016877745.1 | |
| XM_017022257.1 | 24 | 2561 | XP_016877746.1 | |
| XM_017022258.1 | XP_016877747.1 | |||
| XM_017022259.1 | 23 | 2519 | XP_016877748.1 | |
| XM_017022260.1 | 23 | 2495 | XP_016877749.1 | |
| XM_017022261.1 | 24 | 3127 | XP_016877750.1 | |
| XM_017022262.1 | 22 | 2403 | XP_016877751.1 | |
| XM_017022263.1 | 22 | 2426 | XP_016877752.1 | |
| XM_017022264.1 | XP_016877753.1 | |||
| BC012097.1 | 23 | 2516 | AAH12097.1 | |
| BX398277.1 | 1 | 521 | ||
| M99564.1 | 24 | 2530 | AAA36477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039154 | Chr.15:27707535 - 27947424 on Build GRCh38 | Gain |
|
| esv34176 | Chr.15:27461515 - 27774171 on Build GRCh38 | Loss |
|
| nsv1038472 | Chr.15:27647423 - 28256473 on Build GRCh38 | Gain |
|
| nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map