Assay Details
Target Gene Details
Entrez Gene ID: | 80119 |
Gene Name: | PIF1 5'-to-3' DNA helicase |
Gene Aliases: |
C15orf20, PIF |
Location: |
Chr.15:64815630-64827173 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIF1 | NM_001286496.1 | 13 | 2572 | NP_001273425.1 |
| NM_001286497.1 | 13 | 2139 | NP_001273426.1 | |
| NM_001286499.1 | 13 | 2572 | NP_001273428.1 | |
| NM_025049.3 | 13 | 2548 | NP_079325.2 | |
| XM_011522083.2 | 13 | 2664 | XP_011520385.1 | |
| XM_011522084.2 | 13 | 2496 | XP_011520386.1 | |
| AB185926.1 | 13 | 2548 | BAE47454.1 | |
| AB185927.1 | 13 | 2139 | BAE47455.1 | |
| AF108138.1 | 9 | 1652 | AAD44047.1 | |
| AW170361.1 | 1 | 148 | ||
| AY498716.1 | 12 | 2472 | AAS77398.1 | |
| EU084033.1 | 13 | 2543 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv31n68 | Chr.15:64706333 - 64929392 on Build GRCh38 | Loss |
|
| nsv569703 | Chr.15:64756110 - 64930560 on Build GRCh38 | Loss |
|
| esv3636745 | Chr.15:64815095 - 64823057 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map