Assay Details
Target Gene Details
Entrez Gene ID: | 8260 |
Gene Name: | N(alpha)-acetyltransferase 10, NatA catalytic subunit |
Gene Aliases: |
ARD1, ARD1A, ARD1P, DXS707, MCOPS1, NATD, OGDNS, TE2 |
Location: |
Chr.X:153929827-153935154 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA10 | NM_001256119.1 | NP_001243048.1 | ||
| NM_001256120.1 | NP_001243049.1 | |||
| NM_003491.3 | NP_003482.1 | |||
| AK294081.1 | ||||
| AK296593.1 | ||||
| AK315997.1 | ||||
| AY183134.1 | ||||
| AY183135.1 | ||||
| BC000308.2 | AAH00308.1 | |||
| BC019312.1 | AAH19312.1 | |||
| BC063377.1 | AAH63377.1 | |||
| BI192945.1 | ||||
| CR979270.1 | ||||
| X77588.1 | CAA54691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map