Assay Details
Target Gene Details
Entrez Gene ID: | 131 |
Gene Name: | alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide |
Gene Aliases: |
ADH4 |
Location: |
Chr.4:99412261-99435510 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH7 | NM_000673.4 | NP_000664.2 | ||
| NM_001166504.1 | NP_001159976.1 | |||
| AK223472.1 | ||||
| AK301657.1 | ||||
| AK301696.1 | ||||
| AK310811.1 | ||||
| AK312854.1 | ||||
| BC131512.1 | ||||
| BX476080.1 | ||||
| CD014081.1 | ||||
| L33179.1 | AAA59211.1 | |||
| U07821.1 | AAA19002.1 | |||
| U09623.1 | AAA82165.1 | |||
| X76342.1 | CAA53960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv50e213 | Chr.4:99401288 - 99479539 on Build GRCh38 | Gain |
|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| esv3584519 | Chr.4:99402713 - 99556305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map