Assay Details
Target Gene Details
Entrez Gene ID: | 26027 |
Gene Name: | acyl-CoA thioesterase 11 |
Gene Aliases: |
BFIT, STARD14, THEA, THEM1 |
Location: |
Chr.1:54548134-54634744 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT11 | NM_015547.3 | NP_056362.1 | ||
| AB014607.1 | ||||
| AF416921.1 | AAL40937.1 |
Target Gene Details
Entrez Gene ID: | 338094 |
Gene Name: | family with sequence similarity 151 member A |
Gene Aliases: |
C1orf179 |
Location: |
Chr.1:54609177-54623527 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM151A | NM_176782.2 | NP_788954.2 | ||
| XM_006710599.3 | XP_006710662.1 | |||
| AK091901.1 | 1 | 53 | BAC03768.1 | |
| AY358629.1 | AAQ88992.1 | |||
| BC015993.1 | AAH15993.1 | |||
| BC020874.1 | AAH20874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv546269 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain |
|
| esv33550 | Chr.1:54623225 - 54640952 on Build GRCh38 | Gain+Loss |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
| dgv36e212 | Chr.1:54619919 - 54630902 on Build GRCh38 | Loss |
|
| nsv910 | Chr.1:54605786 - 54663322 on Build GRCh38 | Deletion |
|
| nsv829904 | Chr.1:54548613 - 54721384 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map