Assay Details
Target Gene Details
Entrez Gene ID: | 654463 |
Gene Name: | fer-1 like family member 6 |
Gene Aliases: |
C8ORFK23 |
Location: |
Chr.8:123851987-124120061 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FER1L6 | NM_001039112.2 | NP_001034201.2 | ||
| XM_006716618.3 | XP_006716681.1 | |||
| XM_011517231.2 | XP_011515533.1 | |||
| XM_011517232.2 | XP_011515534.1 | |||
| XM_011517233.2 | XP_011515535.1 | |||
| XM_011517235.2 | XP_011515537.1 | |||
| AB196633.1 | BAE53435.1 |
Target Gene Details
Entrez Gene ID: | 439941 |
Gene Name: | FER1L6 antisense RNA 1 |
Gene Aliases: |
C8orf54 |
Location: |
Chr.8:123984138-124040782 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FER1L6-AS1 | NR_040044.1 | |||
| AK093040.1 | BAC04029.1 | |||
| BC137357.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7308n100 | Chr.8:123883276 - 124008232 on Build GRCh38 | Loss |
|
| esv3618675 | Chr.8:123975694 - 124011585 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map