Assay Details
Target Gene Details
Entrez Gene ID: | 55721 |
Gene Name: | IQ motif containing C |
Gene Aliases: |
- |
Location: |
Chr.1:32205635-32208687 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCC | NM_001160042.1 | NP_001153514.1 | ||
| NM_018134.2 | NP_060604.2 | |||
| AK001409.1 | BAA91674.1 | |||
| AK223272.1 | BAD96992.1 | |||
| AK299235.1 | 2 | 426 | ||
| AK307853.1 | ||||
| BC098250.1 | AAH98250.1 | |||
| BC098260.1 | AAH98260.1 | |||
| BC098317.1 | AAH98317.1 | |||
| BC098366.1 | AAH98366.1 | |||
| DA746781.1 |
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM234 | XM_011541795.2 | XP_011540097.1 | ||
| XM_017001814.1 | XP_016857303.1 | |||
| XM_017001815.1 | XP_016857304.1 | |||
| XM_017001817.1 | XP_016857306.1 | |||
| XM_017001818.1 | XP_016857307.1 | |||
| XM_017001819.1 | XP_016857308.1 | |||
| XM_017001821.1 | XP_016857310.1 | |||
| XM_017001827.1 | XP_016857316.1 | |||
| XM_017001828.1 | XP_016857317.1 |
More Information
Set Membership: |
|

Genomic Map