Assay Details
Target Gene Details
Entrez Gene ID: | 6519 |
Gene Name: | solute carrier family 3 member 1 |
Gene Aliases: |
ATR1, CSNU1, D2H, NBAT, RBAT |
Location: |
Chr.2:44275458-44320824 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A1 | NM_000341.3 | 1 | 364 | NP_000332.2 |
| XM_011533047.2 | 1 | 364 | XP_011531349.1 | |
| AB033549.1 | 1 | 345 | BAB16841.1 | |
| AK091932.1 | 2 | 230 | ||
| AK223146.1 | 1 | 326 | BAD96866.1 | |
| AK289636.1 | 1 | 342 | ||
| AK309145.1 | 1 | 358 | ||
| AK313157.1 | 1 | 358 | ||
| BC022386.1 | 1 | 362 | AAH22386.1 | |
| BC093624.1 | 1 | 323 | AAH93624.1 | |
| BC093626.1 | 1 | 318 | AAH93626.1 | |
| DQ023513.1 | 1 | 286 | AAY89644.1 | |
| DQ023515.1 | 1 | 286 | AAY89646.1 | |
| DQ023516.1 | 1 | 286 | AAY89647.1 | |
| DQ023517.1 | 1 | 286 | AAY89648.1 | |
| KU178335.1 | 1 | 286 | ||
| KU178336.1 | 1 | 286 | ||
| L11696.1 | 1 | 329 | AAA81778.1 | |
| M95548.1 | 1 | 330 | AAA35500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map