Assay Details
Target Gene Details
Entrez Gene ID: | 222484 |
Gene Name: | ligand of numb-protein X 2 |
Gene Aliases: |
PDZRN1 |
Location: |
Chr.13:27545913-27621106 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX2 | NM_153371.3 | 5 | 1380 | NP_699202.1 |
| XM_011534995.2 | XP_011533297.1 | |||
| XM_017020434.1 | 5 | 1239 | XP_016875923.1 | |
| AK095319.1 | 5 | 1240 | ||
| AK313394.1 | 5 | 1380 | ||
| BC036755.2 | 5 | 1209 | AAH36755.1 |
More Information
Set Membership: |
|

Genomic Map