Assay Details
Target Gene Details
Entrez Gene ID: | 1384 |
Gene Name: | carnitine O-acetyltransferase |
Gene Aliases: |
CAT1 |
Location: |
Chr.9:129094794-129110791 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRAT | NM_000755.3 | NP_000746.2 | ||
| NM_001257363.1 | NP_001244292.1 | |||
| XM_005251708.3 | XP_005251765.1 | |||
| XM_017014274.1 | XP_016869763.1 | |||
| XM_017014275.1 | XP_016869764.1 | |||
| AK313931.1 | ||||
| BC000723.2 | AAH00723.1 | |||
| BT006801.1 | AAP35447.1 | |||
| X78706.1 | CAA55359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818725 | Chr.9:129026890 - 129176634 on Build GRCh38 | Gain |
|
| nsv831729 | Chr.9:128973042 - 129158330 on Build GRCh38 | Loss |
|
| nsv509323 | Chr.9:128991062 - 129129399 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map