Assay Details
Target Gene Details
Entrez Gene ID: | 79885 |
Gene Name: | histone deacetylase 11 |
Gene Aliases: |
HD11 |
Location: |
Chr.3:13480171-13506424 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC11 | NM_001136041.2 | 10 | 1214 | NP_001129513.1 |
| NM_024827.3 | 10 | 1315 | NP_079103.2 | |
| XM_005265491.1 | 10 | 1280 | XP_005265548.1 | |
| XM_005265492.1 | 7 | 1122 | XP_005265549.1 | |
| XM_006713338.1 | 7 | 1078 | XP_006713401.1 | |
| XM_011534131.1 | 10 | 1403 | XP_011532433.1 | |
| XM_011534132.2 | 7 | 968 | XP_011532434.2 | |
| XM_011534135.1 | 9 | 1122 | XP_011532437.1 | |
| XM_017007234.1 | 7 | 1166 | XP_016862723.1 | |
| AK025426.1 | 10 | 1206 | BAB15127.1 | |
| AK025890.1 | 10 | 1206 | BAB15272.1 | |
| AK293223.1 | 10 | 1147 | ||
| AK299021.1 | 6 | 886 | ||
| AL137362.1 | 7 | 849 | CAB70712.1 | |
| AL834223.1 | 9 | 1530 | CAH56375.1 | |
| BC009676.1 | 9 | 1191 | AAH09676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011766 | Chr.3:13485961 - 14213016 on Build GRCh38 | Gain |
|
| nsv470416 | Chr.3:13492082 - 13575549 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map