Assay Details
Target Gene Details
Entrez Gene ID: | 79676 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:122974703-122980041 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGFOD2 | NM_001304833.1 | NP_001291762.1 | ||
| NM_001304834.1 | NP_001291763.1 | |||
| NM_001304835.1 | NP_001291764.1 | |||
| NM_001304836.1 | NP_001291765.1 | |||
| NM_001304837.1 | NP_001291766.1 | |||
| NM_001304838.1 | NP_001291767.1 | |||
| NM_024623.2 | NP_078899.1 | |||
| AK023553.1 | BAB14610.1 | |||
| AK094820.1 | ||||
| AK095481.1 | ||||
| AK303095.1 | ||||
| BC098119.1 | AAH98119.1 | |||
| BC098293.1 | AAH98293.1 | |||
| BC101940.1 | AAI01941.1 | |||
| BC105637.1 | AAI05638.1 | |||
| BX640675.1 | CAE45807.1 | |||
| BX640734.1 | CAE45849.1 | |||
| DB009535.1 | ||||
| DC404984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv455732 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map