Assay Details
Target Gene Details
Entrez Gene ID: | 79140 |
Gene Name: | coiled-coil domain containing 28B |
Gene Aliases: |
- |
Location: |
Chr.1:32200386-32205390 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC28B | NM_001301011.1 | 1 | 227 | NP_001287940.1 |
| NM_024296.4 | 1 | 227 | NP_077272.2 | |
| XM_017002306.1 | 1 | 227 | XP_016857795.1 | |
| XM_017002308.1 | 1 | 227 | XP_016857797.1 | |
| AK092702.1 | ||||
| AK225113.1 | ||||
| AK291904.1 | 1 | 237 | ||
| BC022848.2 | AAH22848.1 | |||
| DA899719.1 |
More Information
Set Membership: |
|

Genomic Map