Assay Details
Target Gene Details
Entrez Gene ID: | 100526740 |
Gene Name: | ATP5J2-PTCD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:99416739-99466201 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5J2-PTCD1 | NM_001198879.1 | 9 | 2436 | NP_001185808.1 |
| AK295912.1 | 9 | 2399 |
Target Gene Details
Entrez Gene ID: | 26024 |
Gene Name: | pentatricopeptide repeat domain 1 |
Gene Aliases: |
- |
Location: |
Chr.7:99416739-99438839 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTCD1 | NM_015545.3 | 8 | 2403 | NP_056360.2 |
| AB014532.1 | 8 | 2311 | ||
| AB073644.1 | 3 | 685 | ||
| AK001615.1 | 8 | 1796 | ||
| BC003688.1 | 7 | 1916 | AAH03688.2 | |
| BC080580.1 | 8 | 2351 | AAH80580.1 | |
| BC103495.1 | 8 | 2328 | AAI03496.1 | |
| BC103502.1 | 8 | 2328 | AAI03503.1 | |
| BC103661.1 | 6 | 1577 | AAI03662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831069 | Chr.7:99350505 - 99529961 on Build GRCh38 | Loss |
|
| nsv5864 | Chr.7:99411680 - 99443573 on Build GRCh38 | Insertion |
|
| nsv951382 | Chr.7:99416178 - 99434777 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map