Assay Details
Target Gene Details
Entrez Gene ID: | 120425 |
Gene Name: | junction adhesion molecule like |
Gene Aliases: |
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638 |
Location: |
Chr.11:118193727-118225094 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAML | NM_001098526.1 | 3 | 284 | NP_001091996.1 |
| NM_001286570.1 | 2 | 221 | NP_001273499.1 | |
| NM_001286571.1 | 2 | 512 | NP_001273500.1 | |
| NM_153206.2 | 2 | 522 | NP_694938.2 | |
| NR_104479.1 | 2 | 512 | ||
| XM_005271407.3 | 2 | 222 | XP_005271464.1 | |
| XM_017017213.1 | 3 | 209 | XP_016872702.1 | |
| XM_017017214.1 | 3 | 279 | XP_016872703.1 | |
| XM_017017215.1 | 2 | 207 | XP_016872704.1 | |
| AJ515553.2 | 3 | 204 | CAD56620.2 | |
| AK090409.1 | 3 | 184 | BAC03390.1 | |
| AK094399.1 | 2 | 522 | BAC04347.1 | |
| AK298722.1 | 2 | 138 | ||
| AK300851.1 | 3 | 201 | ||
| AK301099.1 | 3 | 201 | ||
| AY093686.1 | 3 | 201 | AAM15730.1 | |
| AY093687.1 | AAM15731.1 | |||
| AY138965.1 | 3 | 185 | AAN52117.1 | |
| AY358362.1 | 3 | 194 | AAQ88728.1 | |
| BC100794.2 | 2 | 211 | AAI00795.1 | |
| BC100795.2 | 2 | 211 | AAI00796.1 | |
| BC100796.1 | 2 | 211 | AAI00797.1 | |
| BC100797.1 | 2 | 211 | AAI00798.1 | |
| DA090367.1 | 2 | 234 | ||
| DC296115.1 | 3 | 284 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511 | Chr.11:118204437 - 118239426 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map