Assay Details
Target Gene Details
Entrez Gene ID: | 57657 |
Gene Name: | hyperpolarization activated cyclic nucleotide gated potassium channel 3 |
Gene Aliases: |
- |
Location: |
Chr.1:155277427-155289848 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCN3 | NM_020897.2 | 1 | 320 | NP_065948.1 |
| NR_073074.1 | 1 | 320 | ||
| XM_011509817.2 | 1 | 320 | XP_011508119.1 | |
| AB040968.2 | BAA96059.2 | |||
| AF488550.1 | 1 | 164 | AAO49470.1 | |
| AK055840.1 | ||||
| AK299333.1 | 1 | 293 | ||
| BC111364.1 | 1 | 274 | AAI11365.1 | |
| DA127524.1 | 1 | 320 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss |
|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv101 | Chr.1:155275103 - 155278531 on Build GRCh38 | Insertion |
|
| nsv436027 | Chr.1:155260761 - 155291532 on Build GRCh38 | Insertion |
|
| esv2758971 | Chr.1:155122502 - 155345016 on Build GRCh38 | Gain |
|
| esv23319 | Chr.1:155257268 - 155292883 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map