Assay Details
Target Gene Details
Entrez Gene ID: | 9016 |
Gene Name: | solute carrier family 25 member 14 |
Gene Aliases: |
BMCP1, UCP5 |
Location: |
Chr.X:130339888-130373361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map