Assay Details
Target Gene Details
Entrez Gene ID: | 127124 |
Gene Name: | ATPase H+ transporting V1 subunit G3 |
Gene Aliases: |
ATP6G3, Vma10 |
Location: |
Chr.1:198523222-198540945 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1G3 | NM_001320218.1 | NP_001307147.1 | ||
| NM_133262.2 | NP_573569.1 | |||
| NM_133326.1 | NP_579872.1 | |||
| XM_006711163.3 | XP_006711226.1 | |||
| XM_011509186.2 | XP_011507488.1 | |||
| XM_011509187.2 | XP_011507489.1 | |||
| AY039760.1 | AAK83465.1 | |||
| BC101129.2 | AAI01130.1 | |||
| BC101130.1 | AAI01131.1 | |||
| BC101131.2 | ||||
| BF509031.1 | ||||
| BX114684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588521 | Chr.1:198316592 - 198769083 on Build GRCh38 | Loss |
|
| nsv548903 | Chr.1:198519360 - 198620446 on Build GRCh38 | Loss |
|
| nsv1004900 | Chr.1:198516035 - 198549828 on Build GRCh38 | Loss |
|
| nsv832259 | Chr.1:198421101 - 198561067 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map