Assay Details
Target Gene Details
Entrez Gene ID: | 23295 |
Gene Name: | mahogunin ring finger 1 |
Gene Aliases: |
RNF156 |
Location: |
Chr.16:4616493-4690974 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGRN1 | NM_001142289.2 | NP_001135761.2 | ||
| NM_001142290.2 | NP_001135762.1 | |||
| NM_001142291.2 | NP_001135763.2 | |||
| NM_015246.3 | NP_056061.1 | |||
| NR_102267.1 | ||||
| XM_005255217.1 | XP_005255274.1 | |||
| XM_005255218.1 | XP_005255275.1 | |||
| XM_005255219.3 | XP_005255276.1 | |||
| XM_005255220.3 | XP_005255277.1 | |||
| XM_005255221.3 | XP_005255278.1 | |||
| XM_011522441.1 | XP_011520743.1 | |||
| XM_017023091.1 | XP_016878580.1 | |||
| XM_017023092.1 | XP_016878581.1 | |||
| XM_017023093.1 | XP_016878582.1 | |||
| AB011116.1 | ||||
| AK299052.1 | ||||
| BC050389.1 | AAH50389.1 | |||
| CN314787.1 | ||||
| DA118421.1 | ||||
| DA590130.1 | ||||
| EF471397.2 | ||||
| EF471398.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv1050237 | Chr.16:4641028 - 4744872 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map