Assay Details
Target Gene Details
Entrez Gene ID: | 27293 |
Gene Name: | sphingomyelin phosphodiesterase acid like 3B |
Gene Aliases: |
ASML3B |
Location: |
Chr.1:27934955-27959157 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPDL3B | NM_001009568.2 | 4 | 701 | NP_001009568.1 |
| NM_001304579.1 | 4 | 772 | NP_001291508.1 | |
| NM_014474.3 | 4 | 701 | NP_055289.2 | |
| XM_011541259.2 | 5 | 791 | XP_011539561.1 | |
| XM_011541260.2 | 5 | 1426 | XP_011539562.1 | |
| AK293489.1 | ||||
| AK293566.1 | 4 | 698 | ||
| AK301349.1 | 4 | 656 | ||
| AK316500.1 | 4 | 627 | ||
| BC014444.1 | 4 | 618 | AAH14444.1 | |
| DC370191.1 | 4 | 700 | ||
| Y08134.1 | 4 | 593 | CAA69328.1 |
More Information
Set Membership: |
|

Genomic Map