Assay Details
Target Gene Details
Entrez Gene ID: | 3071 |
Gene Name: | NCK associated protein 1 like |
Gene Aliases: |
HEM1 |
Location: |
Chr.12:54497711-54543115 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP1L | NM_001184976.1 | NP_001171905.1 | ||
| NM_005337.4 | NP_005328.2 | |||
| AK300783.1 | ||||
| AK314012.1 | ||||
| BC001604.2 | AAH01604.3 | |||
| BC064997.1 | AAH64997.1 | |||
| BC084547.1 | AAH84547.1 | |||
| BC093769.2 | AAH93769.1 | |||
| BC093771.1 | AAH93771.1 | |||
| M58285.1 | AAA35964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508674 | Chr.12:54446126 - 54522212 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map