Assay Details
Target Gene Details
Entrez Gene ID: | 51259 |
Gene Name: | transmembrane protein 216 |
Gene Aliases: |
HSPC244 |
Location: |
Chr.11:61391687-61398863 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM216 | NM_001173990.2 | NP_001167461.1 | ||
| NM_001173991.2 | NP_001167462.1 | |||
| NM_016499.5 | NP_057583.2 | |||
| XM_005274039.3 | XP_005274096.1 | |||
| XM_017017882.1 | XP_016873371.1 | |||
| AF151078.1 | AAF36164.1 | |||
| AK303687.1 | ||||
| BC011010.1 | AAH11010.1 | |||
| BG759319.1 | ||||
| DN998400.1 | ||||
| KU159424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520751 | Chr.11:61389584 - 61415916 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map