Assay Details
Target Gene Details
Entrez Gene ID: | 57409 |
Gene Name: | MIF4G domain containing |
Gene Aliases: |
AD023, MIFD, SLIP1 |
Location: |
Chr.17:75266228-75271292 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map