Assay Details
Target Gene Details
Entrez Gene ID: | 201562 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 2 |
Gene Aliases: |
PTPLB |
Location: |
Chr.3:123491548-123585093 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACD2 | NM_198402.3 | 7 | 798 | NP_940684.1 |
| XM_017005855.1 | 6 | 579 | XP_016861344.1 | |
| AK074605.1 | 3 | 334 | ||
| AK289983.1 | 7 | 798 | ||
| AL832532.1 | 3 | 368 | ||
| AY191814.1 | 7 | 910 | AAP20101.1 | |
| BC049369.1 | 7 | 906 | AAH49369.1 | |
| BC060839.1 | 7 | 800 | AAH60839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829707 | Chr.3:123357546 - 123540363 on Build GRCh38 | Loss |
|
| nsv3978 | Chr.3:123456399 - 123501453 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map