Assay Details
Target Gene Details
Entrez Gene ID: | 3266 |
Gene Name: | ES cell expressed Ras |
Gene Aliases: |
HRAS2, HRASP |
Location: |
Chr.X:48826513-48829869 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERAS | NM_181532.3 | 2 | 588 | NP_853510.1 |
| AB093575.1 | 1 | 759 | BAC76997.1 | |
| BC101642.1 | 1 | 528 | AAI01643.1 | |
| BC101644.1 | 1 | 528 | AAI01645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
| esv3576862 | Chr.X:48755018 - 48837713 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map