Assay Details
Target Gene Details
Entrez Gene ID: | 8001 |
Gene Name: | glycine receptor alpha 3 |
Gene Aliases: |
- |
Location: |
Chr.4:174642011-174829568 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRA3 | NM_001042543.2 | 9 | 3120 | NP_001036008.1 |
| NM_006529.3 | 10 | 3165 | NP_006520.2 | |
| DA409556.1 | 1 | 78 | ||
| U93917.1 | AAC39917.1 |
More Information
Set Membership: |
|

Genomic Map