Assay Details
Target Gene Details
Entrez Gene ID: | 26151 |
Gene Name: | N-acetyltransferase 9 (putative) |
Gene Aliases: |
EBSP, hNATL |
Location: |
Chr.17:74770529-74776425 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT9 | NM_001305077.1 | 7 | 1559 | NP_001292006.1 |
| NM_001305078.1 | 7 | 1558 | NP_001292007.1 | |
| NM_001305079.1 | 7 | 1485 | NP_001292008.1 | |
| NM_001305080.1 | 7 | 1547 | NP_001292009.1 | |
| NM_001305081.1 | 7 | 1556 | NP_001292010.1 | |
| NM_001305082.1 | 7 | 1707 | NP_001292011.1 | |
| NM_001305083.1 | 7 | 1544 | NP_001292012.1 | |
| NM_001305084.1 | 6 | 1443 | NP_001292013.1 | |
| NM_001305085.1 | 6 | 1440 | NP_001292014.1 | |
| NM_001305086.1 | 6 | 1464 | NP_001292015.1 | |
| NM_001305087.1 | 6 | 1463 | NP_001292016.1 | |
| NM_001305088.1 | 6 | 1396 | NP_001292017.1 | |
| NM_015654.4 | 7 | 1562 | NP_056469.2 | |
| NR_130953.1 | 8 | 1566 | ||
| NR_130954.1 | 7 | 1444 | ||
| XM_005257212.4 | 6 | 1401 | XP_005257269.1 | |
| XM_006721813.3 | 6 | 1849 | XP_006721876.1 | |
| XM_017024437.1 | 7 | 1488 | XP_016879926.1 | |
| XM_017024439.1 | 7 | 1452 | XP_016879928.1 | |
| XM_017024441.1 | 5 | 1742 | XP_016879930.1 | |
| AK123115.1 | 7 | 1629 | ||
| AK225602.1 | ||||
| AL050269.1 | 7 | 1439 | CAB43370.1 | |
| AY632082.1 | 6 | 1398 | AAU25819.1 | |
| BC004195.2 | 7 | 1468 | AAH04195.1 | |
| BC004225.2 | 7 | 1457 | AAH04225.1 | |
| HY222854.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map