Assay Details
Target Gene Details
Entrez Gene ID: | 284615 |
Gene Name: | ankyrin repeat domain 34A |
Gene Aliases: |
ANKRD34 |
Location: |
Chr.1:145959441-145964582 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD34A | NM_001039888.3 | 3 | 831 | NP_001034977.1 |
| AK128050.1 | 3 | 831 |
Target Gene Details
Entrez Gene ID: | 84265 |
Gene Name: | RNA polymerase III subunit G like |
Gene Aliases: |
RPC32HOM, flj32422 |
Location: |
Chr.1:145959037-145978848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3GL | XM_005277431.4 | XP_005277488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| nsv8103 | Chr.1:145960053 - 145963570 on Build GRCh38 | Gain |
|
| nsv509457 | Chr.1:145948024 - 149021821 on Build GRCh38 | Insertion |
|
| esv33401 | Chr.1:145961123 - 145963368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map