Assay Details
Target Gene Details
Entrez Gene ID: | 10099 |
Gene Name: | tetraspanin 3 |
Gene Aliases: |
TM4-A, TM4SF8, TSPAN-3 |
Location: |
Chr.15:77044017-77071228 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN3 | NM_001168412.1 | NP_001161884.1 | ||
| NM_005724.5 | NP_005715.1 | |||
| NM_198902.2 | NP_944492.1 | |||
| XM_017021857.1 | XP_016877346.1 | |||
| AF054840.1 | AAC69716.1 | |||
| AF133423.1 | AAF08362.1 | |||
| AK001305.1 | BAA91613.1 | |||
| AK001310.1 | BAA91615.1 | |||
| AK001326.1 | BAA91627.1 | |||
| AK027793.1 | 1 | 117 | ||
| AK075336.1 | ||||
| AK075482.1 | ||||
| AK293677.1 | ||||
| AK298157.1 | ||||
| BC000704.2 | AAH00704.1 | |||
| BC004280.1 | AAH04280.1 | |||
| BC009248.2 | AAH09248.1 | |||
| BC011206.1 | AAH11206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain |
|
| nsv517068 | Chr.15:76962203 - 77088880 on Build GRCh38 | Gain+Loss |
|
| nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map