Assay Details
Target Gene Details
Entrez Gene ID: | 5626 |
Gene Name: | PROP paired-like homeobox 1 |
Gene Aliases: |
CPHD2, PROP-1 |
Location: |
Chr.5:177992235-177996242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROP1 | NM_006261.4 | NP_006252.3 | ||
| AF076215.1 | AAC77454.1 | |||
| BC069076.1 | AAH69076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5844n100 | Chr.5:177883324 - 178184647 on Build GRCh38 | Gain |
|
| nsv509105 | Chr.5:177884347 - 178008855 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map