Assay Details
Target Gene Details
Entrez Gene ID: | 26280 |
Gene Name: | interleukin 1 receptor accessory protein like 2 |
Gene Aliases: |
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1 |
Location: |
Chr.X:104566315-105769729 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RAPL2 | NM_017416.1 | NP_059112.1 | ||
| AF284436.1 | AAG21370.1 | |||
| AJ272208.1 | CAB86868.1 | |||
| AK290196.1 | ||||
| BC104784.1 | AAI04785.1 | |||
| BC104786.1 | AAI04787.1 |
More Information
Set Membership: |
|

Genomic Map