Assay Details
Target Gene Details
Entrez Gene ID: | 641371 |
Gene Name: | acyl-CoA thioesterase 1 |
Gene Aliases: |
ACH2, CTE-1, LACH2 |
Location: |
Chr.14:73493021-73543799 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT1 | XM_017021590.1 | XP_016877079.1 | ||
| XM_017021591.1 | XP_016877080.1 |
Target Gene Details
Entrez Gene ID: | 399671 |
Gene Name: | HEAT repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.14:73478484-73634418 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEATR4 | NM_001220484.1 | NP_001207413.1 | ||
| NM_203309.2 | NP_976054.2 | |||
| XM_006720143.2 | XP_006720206.1 | |||
| XM_011536760.2 | XP_011535062.1 | |||
| XM_011536761.2 | XP_011535063.1 | |||
| XM_017021289.1 | XP_016876778.1 | |||
| AK093515.1 | ||||
| AK302564.1 | ||||
| BC047590.1 | AAH47590.1 | |||
| GU727638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892561 | Chr.14:73365865 - 73620531 on Build GRCh38 | Gain |
|
| nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain |
|
| nsv428295 | Chr.14:73493593 - 73660367 on Build GRCh38 | Gain |
|
| nsv832828 | Chr.14:73498250 - 73664448 on Build GRCh38 | Gain+Loss |
|
| nsv510643 | Chr.14:73483755 - 73582944 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map