Assay Details
Target Gene Details
Entrez Gene ID: | 10747 |
Gene Name: | mannan binding lectin serine peptidase 2 |
Gene Aliases: |
MAP19, MASP-2, MASP1P1, sMAP |
Location: |
Chr.1:11026523-11047239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASP2 | NM_006610.3 | NP_006601.2 | ||
| NM_139208.2 | NP_631947.1 | |||
| XM_017000097.1 | XP_016855586.1 | |||
| AB008047.1 | BAA78616.1 | |||
| AK290823.1 | ||||
| X98400.1 | CAA67050.1 | |||
| Y09926.1 | CAA71059.1 | |||
| Y18281.1 | CAB50728.1 | |||
| Y18282.1 | CAB50729.1 | |||
| Y18283.1 | CAB50730.1 | |||
| Y18284.1 | 4 | 1006 | CAB50731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832425 | Chr.1:10843788 - 11062918 on Build GRCh38 | Loss |
|
| esv2660954 | Chr.1:11037555 - 11046258 on Build GRCh38 | Deletion |
|
| esv3585212 | Chr.1:11037585 - 11046365 on Build GRCh38 | Loss |
|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
| nsv953456 | Chr.1:11038744 - 11060143 on Build GRCh38 | Deletion |
|
| esv25881 | Chr.1:11037505 - 11045994 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map