Assay Details
Target Gene Details
Entrez Gene ID: | 29964 |
Gene Name: | prickle planar cell polarity protein 4 |
Gene Aliases: |
C6orf49, OBTP, OEBT, TOMM6 |
Location: |
Chr.6:41780762-41787372 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE4 | NM_013397.5 | 7 | 902 | NP_037529.3 |
| BC110459.1 | 5 | 1354 | AAI10460.1 | |
| DA917216.1 | 3 | 516 |
More Information
Set Membership: |
|

Genomic Map