Assay Details
Target Gene Details
Entrez Gene ID: | 100527960 |
Gene Name: | MROH7-TTC4 readthrough (NMD candidate) |
Gene Aliases: |
HEATR8-TTC4 |
Location: |
Chr.1:54641740-54742657 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MROH7-TTC4 | NR_037639.2 | 33 | 5437 | |
| NR_037640.2 | 33 | 5304 | ||
| NR_037641.2 | 33 | 5083 | ||
| AK127008.1 | 17 | 2938 | ||
| BX648088.1 | 33 | 5243 | ||
| CR749830.1 | 33 | 5389 | CAH18687.1 |
Target Gene Details
Entrez Gene ID: | 7268 |
Gene Name: | tetratricopeptide repeat domain 4 |
Gene Aliases: |
- |
Location: |
Chr.1:54715822-54742657 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC4 | NM_001291333.1 | 8 | 1049 | NP_001278262.1 |
| NM_004623.4 | 10 | 1346 | NP_004614.3 | |
| AF063602.1 | 10 | 1308 | AAG43161.1 | |
| AF073887.1 | 10 | 1291 | AAD19853.1 | |
| AK025524.1 | 10 | 1346 | ||
| AK090964.1 | 10 | 1426 | ||
| AK123924.1 | 2 | 1174 | BAC85726.1 | |
| AK222562.1 | 10 | 1299 | BAD96282.1 | |
| AK292426.1 | 10 | 1307 | ||
| BC001276.1 | 10 | 1285 | AAH01276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004778 | Chr.1:54635180 - 54764639 on Build GRCh38 | Gain |
|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |
|
| nsv546269 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain |
|
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|

Genomic Map